Find methods for your needs
Separation of related impurities and intermediate stages of Guanine
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied to the separation of related impurities and Intermediate stage separation of Guanine. The separation was performed on a Thermo Scientific Acclaim Mixed Mode WCX-1 column with a UV detection at 254 nm. With a run time of 15 minutes, the method allows easy separation of intermediate stages and related impurities of Guanine.| Market: | Pharma |
| Keywords: | HPLC, UV detection, Guanine, 2,4-Diamino-6-hydroxypyrimidin, 2,4-diamino-5-isonitroso-6-hydroxypyrimidine(DNHP), 2,4,5-triamino-6-hydroxypyrimidine sulphate(THP), Acclaim Mixed Mode WCX-1 |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.