Find methods for your needs
Separation of process intermediate stages of Glutamine.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of Glutamine intermediates The separation was performed on a Thermo Scientific Acclaim Polar Advantage HPLC column. There are 3 different stages which are highly polar, mid polar and non polar in nature and need to be separated, these stages are termed as Stage-IC, Stage-II-B and Stage-III Stage-I-C, Stage-II B and Stage –III are well separated and are easily detected up to 0.15 ppm when spiked with Stage-III.| Market: | Pharma |
| Keywords: | HPLC, UV detection, Glutamine, Acclaim Polar Advantage C16, Intermediate stages |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.