Find methods for your needs
Fast analysis of a fexofenadine tablet using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of a fexofenadine tablet. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Fexofenadine, HPLC, Hypersil GOLD, Tablet, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.