Find methods for your needs
AN1081: Automated Solid-Phase Extraction of Triclosan in Tap Water Using Hydrophilic Reversed-Phase Cartridges and HPLC with UV Detection
Description
In this application note, triclosan-contaminated samples of tap water were simulated by adding known amounts of triclosan-containing liquid hand soap to Sunnyvale tap water. The Dionex AutoTrace 280 instrument along with the high-surface-area, hydrophilic, reversed-phase cartridges were used to concentrate triclosan. The contents of the cartridge eluent were then separated on a reversed phase column using HPLC with UV detection.Market: | Environmental |
Keywords: | antifungal, Cosmetics, Triclosan, Toothpaste, Dionex AutoTrace 280, antimicrobial agent, Liquid hand soap, SolEx SPE HRPHS cartridges, Body wash |
Matrix: | Drinking water |
Author: | Jing Hong and Rosanne Slingsby |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.