Find methods for your needs
Determination of chloramphenicol using the USP method and a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of chloramphenicol using the USP method. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column in less than 5 minutes, with UV detection at 280 nm.| Market: | Pharma |
| Keywords: | Antibiotic, Chloramphenicol, HPLC, Syncronis C18, USP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.