Find methods for your needs
Improved analysis of allantoin using a Thermo Scientific Sycnronis HILIC HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of allanotin. The separation is performed on a Thermo Scientific Syncronis HILIC HPLC column, with UV detection at 210 nm.
Market: Pharma
Keywords: Allantoin, HILIC, HPLC, Syncronis HILIC, UV detection
Matrix: N/A
Uploaded on 8/20/2012.
For Research Use Only. Not for use in diagnostic procedures.