Find methods for your needs
TN148: Automated Extraction and Determination of Human Hormones in Drinking Water Using Solid-Phase Extraction and HPLC with UV Detection
Description
This Technical Note describes the use of high-surface-area solid phase extraction (SPE) to extract seven human hormones from drinking water using the Dionex AutoTrace 280 SPE instrument. Extracts were subsequently quantified using high-performance liquid chromatography (HPLC) to determine the recovery.| Market: | Environmental |
| Keywords: | Acclaim RSLC 120 C18, SPE, UltiMate 3000, Estrogen, Drinking water, Endocrine disruptor, androgen, Dionex AutoTrace 280, EPA Method 539, SolEx HRPHS, Sex hormones, hormone replacment |
| Matrix: | Drinking water |
| Author: | Carl Fisher and Linda Lopez |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.