Find methods for your needs
Improved analysis of urea herbicides using a Thermo Scientific Syncronis C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of urea herbicides. The separation is performed on a Thermo Scientific Syncronis C8 HPLC column, with UV detection at 240 nm.| Market: | Environmental |
| Keywords: | Chlorotoluron, Diuron, HPLC, Linuron, Metoxuron, Monuron, Syncronis C8, Tebuthiuron, Urea herbicides, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.