Find methods for your needs
Dionex IonPac AS11-HC-4µm Column Performance Test Using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. At least three injections should be made. This record provides an eWorkflow for executing the QAR method on an ICS-5000 system.| Market: | Environmental; Food and Beverage |
| Keywords: | AERS 500, Ion Chromatography (IC), IonPac AS11-HC-4um column, suppressed conductivity detection, EGC 500 KOH, Quality Assurance Report (QAR) |
| Matrix: | <DI water> |
| Author: | Beibei Huang; Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.