Find methods for your needs
Non-targeted Screening and Accurate Mass Confirmation of 510 Pesticides on the High Resolution Exactive Benchtop LC/MS Orbitrap Mass Spectrometer
Description
As agricultural trade grows and food safety concerns mount, stricter pesticide regulations are being enforced around the world. Increased pesticide testing and reductions in maximum permissible residue levels have driven demand for fast, sensitive and cost-effective analytical methods for high-throughput screening of multi-class pesticides in food. Detection of 510 pesticides at low ppb levels was achieved within 12 minutes using the Thermo Scientific Exactive benchtop LC/MS system powered by Orbitrap technology.| Market: | Food and Beverage |
| Keywords: | High resolution, Pesticide Analysis, Orbitrap Technology, Exactive, High Mass Accuracy |
| Matrix: | Spinach |
| Author: | Allen Zhang, James S. Chang, Christine Gu, Mark Sanders |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.