Find methods for your needs
Validated Method for the Analysis of Lipophilic Marine Biotoxins in Bivalves by Liquid Chromatography-High-Resolution Mass Spectrometry
Description
A harmful algal bloom is a serious environmental problem that results when the population of microscopic algae in a water system rapidly increases. The algae have the ability to produce marine biotoxins (MBTXs), which are dangerous to humans and other aquatic life. These biotoxins can accumulate in filter-feeding bivalve mollusks and, in turn, pose significant food safety risks to humans who ingest the shellfish, including gastrointestinal illnesses or neurological disorders. We describe a UHPLC with HRMS/MS method that delivers excellent results in the quantitative analysis of MBTXs.| Market: | Environmental |
| Keywords: | LC-MS, Accela UHPLC, Q Exactive MS, Marine biotoxins, QuEChERS sample prep |
| Matrix: | Mussel |
| Author: | Complete method: Rúbies, A. et al (J. Chrom. A, 2015 Mar 20;1386:62-73) |
| Affiliation: | Laboratori de l’Agència de Salut Pública de Barcelona-Spain |
For Research Use Only. Not for use in diagnostic procedures.