Find methods for your needs
Metabolomic Analysis of Green and Black Tea Extracts Using an LTQ Orbitrap XL Hybrid Linear Ion Trap Mass Spectrometer
Description
We demonstrate a complete analytical metabolomic workflow including (1) data acquisition using a high resolution accurate mass instrument that is equipped with a Higher Energy Collisional Dissociation (HCD) cell and coupled to a high pressure LC (Figure 1), (2) metabolite differential abundance analysis, and (3) structural elucidation of relevant metabolites using accurate mass and HCD fragmentation information to highlight the component differences between green and black tea.| Market: | Metabolomics; Food and Beverage |
| Keywords: | Mass Frontier Software, SIEVE Software, Accela High Speed UHPLC, Higher Energy Collisional Dissociation (HCD), Natural Product Analysis |
| Matrix: | Tea |
| Author: | Donna L. Wilson and Charles Yang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.