Find methods for your needs
Detection of Mycotoxins in Corn Meal Extract Using Automated Online Sample Preparation with LC-MS/MS
Description
Methyl 2-benzimidazole carbamate, most commonly known as carbendazim, is a widely used broad-spectrum benzimidazole fungicide and a decomposition product of benomyl. Carbendazim is used to control plant diseases in cereals and fruit, including citrus, bananas, strawberries, pineapples, and pome fruits. We describe a method that is an adaptation of an existing online, multi-residue pesticide method (Thermo Scientific Method 522133) proven and verified specially for the actual carbendazim contamination issue of orange juices in the US.| Market: | Food and Beverage |
| Keywords: | Mycotoxins, Food Safety, TurboFlow Technology, TSQ Vantage Triple Quad MS, Aria Operating Software |
| Matrix: | Juice |
| Author: | Yang Shi, Catherine Lafontaine, François Espourteille |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.