Find methods for your needs
Analysis of Triazine Herbicides in Drinking Water Using LC-MS/MS and TraceFinder Software
Description
We demonstrate the capabilities of Thermo Scientific TraceFinder software with a mixture of triazine compounds spiked into drinking water samples was analyzed. Using direct injections of 20 mL samples (with on-line preconcentration), low- and sub-pg/mL (ppt) levels were detected. The ability to analyze these drinking water samples with on-line preconcentration saves considerable time and expense compared to solid phase extraction techniques.Market: | Environmental |
Keywords: | Herbicides, environmental, Water analysis |
Matrix: | Water |
Author: | Jonathan R. Beck, Jamie K. Humphries, Louis Maljers, Kristi Akervik, Charles Yang, Dipankar Ghosh |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.