Find methods for your needs
Validated Method for the Determination of Phenylbutazone in Horse Meat with LC-MS/MS
Description
Phenylbutazone (bute, PBT) is one of the most widely used (non-steroidal) anti-inflammatory (NSAiD) and painkiller drug for dogs and horses. Horse meat is still widely used for human consumption in some European countries. For those animals marked for human consumption, a limited number of medicines can be administered. A newly developed LC-MS/MS method for the determination of PBT in horse meat with LC-MS/MS is described here. The method was validated in-house and its applicability for routine testing was tested following the European legislation requirements.| Market: | Food and Beverage |
| Keywords: | NSAID, Adulteration, TSQ Vantage, Horse Meat, In-house Validation, Phenylbutazone, Triple Quadrupole Mass Spec |
| Matrix: | Horse meat |
| Author: | Katerina Bousova, Laszlo Hollosi, Ebru Ates, Michal Godula |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.