Find methods for your needs
Analysis of Triazine Pesticides in Drinking Water Using LC-MS/MS (EPA Method 536.0)
Description
The US EPA has recently issued a draft form of a proposed method for the analysis of triazine compounds in drinking water. This method uses a simple method to directly analyze triazine compounds using LC-MS/MS without requiring any solid phase extraction (SPE) or other lengthy sample preparation steps. This application note demonstrates the analysis of these compounds over the concentration range 0.25 – 5.0 ng/mL (ppb) using the Thermo Scientific TSQ Quantum Access triple stage quadrupole mass spectrometer and the Thermo Scientific Accela HPLC system.| Market: | Environmental |
| Keywords: | Herbicides, Triazines, Drinking water analysis |
| Matrix: | Water |
| Author: | Jonathan R. Beck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.