Find methods for your needs
Quantitative and Qualitative Confirmation of Pesticides in Beet Extract Using a Hybrid Quadrupole-Orbitrap Mass Spectrometer
Description
We describe a method for the analysis of pesticides, showing the utility of a full-scan data-dependent MS/MS workflow to achieve regulatory levels while providing a complete targeted and screening analysis using a high-resolution, accurate mass (HRAM) spectral library for identification and confirmation.| Market: | Food and Beverage |
| Keywords: | High resolution, Pesticides, Accurate mass, Quantitation, Target screening, Unknown screening, Retrospective data analysis, LCMSMS HRAM |
| Matrix: | Beets |
| Author: | Charles Yang, Olaf Scheibner, Dipankar Ghosh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.