Description
本文参照ASTM D7328对变性乙醇燃料样品进行前处理后,选用高容量 IonPac AS22高效阴离子交换分离柱完成了样品中痕量游离氯化物和硫酸盐及 总硫的含量测定。方法选择性较好,氯离子和硫酸盐的分离不受样品基质的 影响,其定量结果更加准确。| Market: | Petrochem, Oil and Gas |
| Keywords: | 离子色谱;电导;IonPac AS22;变性乙醇燃料;氯化物;硫酸根 |
| Matrix: | 变性乙醇燃料 |
| Author: | 郑洪国 |
For Research Use Only. Not for use in diagnostic procedures.