Description
A method for the purity analysis of mannose-6-phosphate using an isocratic HILIC elution is described capturing free phosphate, a related impurity, and the sodium ion using the Acclaim Trinity P2 column and the Corona Veo detector. A high injection volume (40 µL) was used on a sample of 1 mg/mL of mannose-6-phosphate to show the free phosphate. A higher concentration and lower injection volume can also be used.| Market: | Other |
| Keywords: | Mannose 6-phosphate |
| Matrix: | Water |
| Author: | Marc Plante |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.