Find methods for your needs
TN73: Determination of PFOA and PFOS in Water Samples Using On-Line Sample Concentration, Reversed-Phase Liquid Chromatography and Suppressed Conductivity Detection
Description
This technical note describes a methodology to determine Perfluorooctanoic Acid (PFOA) and Perfluorooctanesulfonic Acid (PFOS) in water samples using on-line sample concentration, reversed-phase HPLC separation, and suppressed conductivity detection. Instrument setup, sample concentration, chromatographic method, calibration, and limits of detection are detailed.| Market: | Environmental |
| Keywords: | EPA, Acclaim PA2, PFOA, PFOS, Perfluorinated acids, Teflon, TN73, PFAS, PFCA |
| Matrix: | Drinking water |
| Author: | Mark L. Tracy |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.