Find methods for your needs
USP Monograph: Content of flavonol glycosides in ginkgo tablets using C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the content of flavonol glycosides in ginkgo tablets. The separation was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the USP 38 monograph.The system suitability parameters met the criteria stated in the USP.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, USP monograph, UV detection |
| Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.