Find methods for your needs
TN23: Ion Chromatography of Lanthanide Metals (Part 2/3: Anion Exchange Separation of Lanthanide Metals)
Description
The separator column used for this method is the IonPac CS5.The separation is achieved using opposing linear gradients of a-hydroxyisobutyric acid (HIBA) oxalic acid and diglycolic acid.Market: | Chemical |
Keywords: | Anion Exchange, Ion Chromatography, lanthanide metals, PAR Reagent |
Matrix: | Water |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.