Find methods for your needs
AB141: Fast Determination of Inorganic Anions in Municipal Drinking Water Using Capillary Ion Chromatography.
Description
The determination of common inorganic anions in drinking water is one of the most important applications of ion chromatography (IC) worldwide. This application brief describes the fast determination of inorganic anions in drinking water samples with the Dionex ICS-5000 capillary system using the Dionex IonSwift MAX-100 column. The high pressure capability of this system facilitates higher sample throughput by simply increasing the flow rate of analysis.| Market: | Water Analysis |
| Keywords: | HPIC, fluoride, Nitrate, Nitrite, Chloride, phosphate, Sulfate, capillary, IonSwift MAX-100, RFIC, Drinking water, Bromide, Carbonate, Municipal |
| Matrix: | Drinking water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.