Description
Saccharin decreases the tensile strength of electrolytic nickel deposits and can, with increasing concentrations, cause compressive stress. Determination of saccharin by ion chromatography is faster and has fewer interferences than classical titrimetric methods of analysis.| Market: | Other |
| Keywords: | Acetonitrile, brightener, Nickel, OmniPac PAX-500, plating bath, Saccharin, sulfuric acid, electrolytic |
| Matrix: | Nickel sulfate |
| Affiliation: | Dionex Corporation |
For Research Use Only. Not for use in diagnostic procedures.