Find methods for your needs
Related substances analysis of ranolazine by reversed-phase HPLC-UV
Description
The impurity analysis of ranolazine was performed using a Thermo Scientific Dionex UltiMate 3000 HPLC system and a Thermo Scientific Accucore C18 HPLC column. The method was performed in 40 minutes with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | HPLC, UV detection, Ranolazine |
| Matrix: | Formulation |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.