Find methods for your needs
EP 8.0 monograph - impurity determination of ofloxacin using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of ofloxacin and its impurities. The separation was performed on a Thermo Scientific Syncronis C18 HPLC column using the method described in the EP 8.0 monograph. The results obtained for resolution exceeded the criteria stated in the EP. Resolution between impurity E and ofloxacin was 3.3 (EP criteria of not less than 2.0).| Market: | Pharma |
| Keywords: | HPLC, Ofloxacin, Syncronis C18, UV detection, EP 8.0 monograph |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.