Find methods for your needs
USP 38 monograph - impurity determination of ceftriaxone sodium using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of ceftriaxone sodium and its impurities. The separation was performed on a Thermo Scientific Syncronis C18 HPLC column using the method described in the USP 38 monograph. The results obtained for resolution and signal to noise ratio exceeded the criteria stated in the USP. Resolution between ceftriaxone sodium and ceftriaxone sodium E isomer peak was 9.99 (USP criteria of not less than 3.0). Signal to noise ratio of ceftriaxone sodium peak in standard solution was 553 (USP criteria not less than 10).| Market: | Pharma |
| Keywords: | HPLC, Syncronis C18, UV detection, USP 38 monograph, Ceftriaxone sodium |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.