Find methods for your needs
AN1091: Determination of Uronic Acids and Wood Sugars in Wood-Based Hydrolysates: Uronic Acid Method
Description
Wood, grass, and other lignocellulosic biomass are emerging as the preferred sources for biofuel and biochemical production. Compositional analysis of plant derived material is crucial for maximizing the yield of biofuel production. This record shows an HPAE-PAD Method that uses the Dionex CarboPac PA200 column to determine uronic acids in wood hydrolysates.| Market: | Other |
| Keywords: | biofuel, Electrochemical detection, CarboPac PA200, Galacturonic Acid, glucuronic acid, wood hydrolysate, HPAE-PAD, TN21, TN71, TN110, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Wood |
| Author: | Lipika Basumallick and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.