Description
Choline and acetylcholine are quaternary ammonium cations which are noted for their activities as neurotransmitters. This is an older application note that shows these compounds can be determined by IC. Today we would probably use an IonPac CS19 column as shown in Application Update 189.Market: | Pharma |
Keywords: | Acetylcholine, choline, Dietary supplements, Neurotransmitters, Quaternary amine, pill, OmniPac PCX-100 |
Matrix: | Pill; Tablet |
For Research Use Only. Not for use in diagnostic procedures.