Find methods for your needs
AU165: Separation of Chromium (III) and Chromium (VI) by Ion Chromatography
Description
This application update demonstrates separation and detection of Cr(III) and Cr(VI) using a Dionex IonPac CS5A column and absorbance detection. This method also describes a sample preparation technique that can be used to estimate the concentrations of Cr(III) and Cr(VI) in soil and wastewater samples.| Market: | Environmental |
| Keywords: | Ion Chromatography (IC), Cr(III), Cr(VI), Dionex IonPac CS5 column, soil, Wastewater |
| Matrix: | Soil, Water |
| Author: | T.Phesatcha, W. Worawirunwong, Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.