Find methods for your needs
AU168: Determination of Transition Metals in Complex Matrices Be the first to rate this application
transition metals complex matrices
Description
Chelation ion chromatography facilitates the determination of low concentrations (μg/L and lower) of transition metals in samples including seawater, brines, estuarine waters, and a variety of biological samples. Here, the authors simplify the system configuration described in Technical Note 25, using an ICS-3000 system. The separation has also been updated to include the IonPac CS5A/CG5A column set, which demonstrates improved selectivity and peak efficiency for separation of transition metals compared to the CS5.
Market: Water Analysis
Keywords: copper, Iron, Zinc, Chelation Ion Chromatography, nickel, Cadmium, Cobalt, seawater, manganese, IonPac CS5A, Fe3+, Cu2+, Ni2+, Zn2+, Co2+, Cd2+, Mn2+, Alkaline earth metals, Postcolumn derivatization
Matrix: Sea water
Author: Chanita Chantarasukon, Weerapong Worawirunwong, Jeff Rohrer, Archava Siriraks
Affiliation: Thermo Fisher Scientific
Uploaded on 8/22/2015.
For Research Use Only. Not for use in diagnostic procedures.