Find methods for your needs
Separation and detection of 15 triazine herbicides by GC-MS
Description
The Thermo Scientific Trace 1310 ISQ system is applied for the direct injection and analysis of triazine herbicide samples. Separation is performed on a Thermo Scientific TraceGOLD TG-5SilMS column with MS SIM detection.| Market: | Environmental |
| Keywords: | GC-MS, Herbicides, Triazines, MS detection, TraceGOLD TG5SilMS |
| Matrix: | N/A |
| Author: | Richard Law |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.