Find methods for your needs
AN133: Determination of Inorganic Anions in Drinking Water by Ion Chromatography - A
Description
The determination of common inorganic anions in drinking water is one of the most important applications of ion chromatography (IC) worldwide. Ion chromatography has been approved for compliance monitoring of these common inorganic anions in U.S. drinking water since the mid-1980s, as described in U.S. EPA Method 300.0. This application note describes the determination of inorganic anions in drinking water and other environmental waters using an IonPac AS4A-SC column and conditions that are consistent with those in U.S. EPA Method 300.0.Market: | Environmental |
Keywords: | bromide, chloride, drinking water, EPA Method 300, EPA Method 300.0, EPA Method 300.0 Part A, fluoride, IonPac AS4A-SC column, nitrate, nitrite, phosphate, sulfate |
Matrix: | Drinking water |
Author: | Peter Jackson |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.