Find methods for your needs
AU126: Determination of Diethanolamine and Triethanolamine in Surface Finishing, Wastewater and Scrubber Solutions
Description
Alkanolamines are important in the chemical, pharmaceutical, and petroleum industries. Their analysis is also important in metal surface finishing and in wastewater effluents. Ion chromatography (IC) works particularly well for separations of alkanolamines. This application update describes the use of the OmniPac PAX-500 column with Pulsed Amperometric Detection (PAD) to measure alkanolamines.| Market: | Petrochem, Oil and Gas |
| Keywords: | Diethanolamine, OmniPac PAX-500, TEA, PAD, DEA, Alkanolamines, triethanolamine, Scrubber solution, Etch solution, Oil Refinery, Pond water |
| Matrix: | Scrubber solution |
| Author: | Edward Kaiser |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.