Find methods for your needs
EP 8.0 monograph - impurities test for piracetam using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of piracetam and its impurities. The separation was performed on a Thermo Scientific Syncronis C18 HPLC column using the method described in the USP monograph. The results obtained for resolution and asymmetry exceeded the criteria stated in the USP. Resolution between piracetam and impurity A was 4.8 (USP criteria of not less than 3.0). Asymmetry of piracetam was 1.2 (USP criteria of not more than 2.0).| Market: | Pharma |
| Keywords: | HPLC, Syncronis C18, Piracetam, UV detection, EP 8.0 monograph |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.