Find methods for your needs
Fast determination of retinyl palmitate and retinyl acetate
Description
The Thermo Scientific HPLC system is applied for the analysis of retinyl palmitate and retinyl acetate. The separation is performed on a Thermo Scientific Acclaim HILIC-10 column, with UV detection at 285 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim HILIC-10, HILIC, HPLC, Retinyl acetate, Retinyl palmitate, UV detection, Vitamin A |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.