Find methods for your needs
A simple gradient method for the analysis of resveratrol in red wine
Description
The Thermo Scientific UltiMate 3000 HPLC system is applied for the analysis of resveratrol in red wine. The separation is performed on a Thermo Scientific Acclaim 120 C18 column, with UV detection at 310 nm.| Market: | Food and Beverage |
| Keywords: | Acclaim 120 C18, HPLC, Red wine, Resveratrol, UV detection |
| Matrix: | Red wine |
For Research Use Only. Not for use in diagnostic procedures.