Find methods for your needs
Comparison of the analysis of a peptide mix with formic acid mobile phase additive
Description
The Thermo Scientific HPLC system is applied for the analysis of a peptide mix. The separation is performed on both a Thermo Scientific Acclaim 300 C18 HPLC column (chromatogram A) and Thermo Scientific PolarAdvantage II (PA2) HPLC column (chromatogram B). This comparison shows that when formic acid is used instead of TFA for peptide analysis the Acclaim PA2 provides a complementary selectivity compared to the C18 phase.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, Acclaim PolarAdvantage II (PA2), HPLC, Peptide mix, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.