Find methods for your needs
Quick determination of miconazole in athlete's foot ointment
Description
The Thermo Scientific HPLC system is applied for the analysis of miconazole in athlete's foot ointment. The separation is performed on a Thermo Scientific Acclaim 120 C18 column, with UV detection at 225 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Athlete's foot ointment, Benzoic acid, HPLC, Miconazole, UV detection |
| Matrix: | Athlete's foot ointment |
For Research Use Only. Not for use in diagnostic procedures.