Description
The Thermo Scientific HPLC system is applied for the analysis of hydrocortisone in skin ointment. The separation is performed on a Thermo Scientific Acclaim 120 C8 column, with UV detection at 245 nm.| Market: | Pharma |
| Keywords: | Acclaim 120 C8, HPLC, Hydrocortisone, Methyl paraben, Propyl paraben, Skin ointment, UV detection |
| Matrix: | Skin ointment |
For Research Use Only. Not for use in diagnostic procedures.