Find methods for your needs
Determination of polyacrylic acid in boiler water by HPLC-CAD
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of polyacrylic acid in boiler water. The separation is performed on a Thermo Scientific Acclaim SEC-1000 column, with charged aerosol detection.| Market: | Chemical |
| Keywords: | Acclaim SEC-1000, Boiler water, Charged aerosol detection, HPLC, Polyacrylic acid (PAA), SEC |
| Matrix: | Boiler water |
| Author: | Mark Tracy, Xiaodong Liu, Ian Acworth |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.