Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of ursolic acid in apple peel. The separation is performed on a Thermo Scientific Acclaim C30 column, with UV detection at 210 nm. Chromatogram A is extracted Fuji apple, chromatogram B is extracted red delicious apple and chromatogram C is standards at 100 μg/mL.| Market: | Food and Beverage |
| Keywords: | Acclaim C30, Apple peel, HPLC, Oleanolic acid, Ursolic acid, UV detection |
| Matrix: | Apple peel |
For Research Use Only. Not for use in diagnostic procedures.