Find methods for your needs
USP 38 monograph: impurity determination of carbamazepine using a cyano HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of carbamazepine and its impurities. The separation was performed on a Thermo Scientific Hypersil Gold CN HPLC column using the method described in the USP 38 monograph. The results obtained for resolution exceeded the criteria stated in the USP. Resolution between carbamazepine related compound A and carbamazepine was 4.00 (USP criteria of not less than 1.70).| Market: | Pharma |
| Keywords: | Carbamazepine, HPLC, Hypersil GOLD CN, UV detection, USP 38 monograph |
| Matrix: | N/A |
| Author: | Manish singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.