Find methods for your needs
Simultaneous analysis of sodium and glutamate in food products. Be the first to rate this application
simultaneous analysis sodium glutamate food products
Description
A gradient HPLC method was developed for the direct analysis of sodium and glutamate present in a popular food product from India called Maggi 2-Minute Noodles. Chromatographic separation was performed using the mixed mode Acclaim Trinity P1 column. One advantage in using this multi-modal column is that several ions such as sodium and chloride can be simultaneously measured along with glutamate present in the Maggi spice mixture. A simple direct method for the analysis of various amino acids is possible using the universal charged aerosol detector (CAD) as previously published.
Market: Food and Beverage
Keywords: amino acid, food, HPLC CAD
Matrix: Food
Author: Bruce Bailey
Affiliation: Thermo Scientific, CMD
Uploaded on 6/22/2015.
For Research Use Only. Not for use in diagnostic procedures.