Description
The Thermo Scientific HPLC system is applied for the analysis of quaternary ammonium salts. The separation is performed on a Thermo Scientific Hypercarb column, with UV detection at 295 and 245 nm.| Market: | Environmental |
| Keywords: | Diquat, HPLC, Hypercarb, Paraquat, Quaternary ammonium salts, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.