Find methods for your needs
AN43099: IC-ICP-MS speciation analysis of As in apple juice
Description
Inorganic and organic arsenic compounds have very different toxicities, and therefore speciation analysis is needed to assess the potential health risk posed by the food product. Here two inorganic and four organic arsenic species are determined in an apple juice sample using IC coupled to iCAP Q ICP-MS. Arsenic species are first separated by IC using an ammonium carbonate gradient on an IonPac AS7 column. Then the eluting species are detected and quantified by the ICP-MS down to ng/g concentrations. Arsenic speciation is also demonstrated in organic brown rice syrup (OBRS) in AN43126.| Market: | Food and Beverage |
| Keywords: | MMA, ion chromatography, apple juice, arsenate, arsenic, arsenite, As, IC, IC-ICP, IC-ICP-MS, ICP-MS, IonPac AS7 column, speciation, Arsenobetaine, AsB, DMA, Arsenocholine, Monomethylarsonic, Dimethylarsinic, As (III) |
| Matrix: | Apple juice |
| Author: | D. Kutscher, S. McSheehy, J. Wills, D. Jensen |
For Research Use Only. Not for use in diagnostic procedures.