Description
The Thermo Scientific Hypersil GOLD Silica column was used to separate seven steroids using a normal-phase HPLC method with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | 11-a-Hydroxyprogesterone, 17-a-Hydroxyprogesterone, 21-Hydroxyprogesterone-21-acetate, Corticosterone, Cortisone, HPLC, Hydrocortisone, Hypersil GOLD Silica, Progesterone, Steroids, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.