Find methods for your needs
Simple gradient method for the analysis of a protein mixture
Description
The Thermo Scientific HPLC system is applied for the analysis of a protein mixture. The separation is performed on a Thermo Scientific Acclaim 300 C18 column, with UV detection at 214 nm.| Market: | BioPharma |
| Keywords: | Acclaim 300 C18, Carbonic anhydrase, HPLC, Impurities, Lysozyme, Myoglobin, Protein, RNAse A, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.