Find methods for your needs
Improved gradient separation of propafenone and related substances
Description
The Thermo Scientific HPLC system is applied for the analysis of propafenone and related substances. The separation is performed using a Thermo Scientific Acclaim PolarAdvantage column, with UV detection at 249 nm.| Market: | Pharma |
| Keywords: | Acclaim PolarAdvantage, Chlorohydrine, Dimer, Epoxyefenone, HBAH, HPLC, Propafenone, Tertiary amine, UV detection |
For Research Use Only. Not for use in diagnostic procedures.