Find methods for your needs
Enhanced impurity profile of 2-(N,N-dimethylamino)ethyl acrylate (DMAEA)
Description
The Thermo Scientific HPLC system is applied for the impurity profile of 2-(N,N-dimethylamino)ethyl acrylate. The separation is performed using a Thermo Scientific Acclaim PolarAdvantage column, with UV detection at 210 nm. The three chromatograms represent three different batches of DMAEA.| Market: | Material Science |
| Keywords: | 2-(NN-dimethylamino)ethyl acrylate, Acclaim PolarAdvantage, DMAEA, HPLC, Impurity profile, UV detection, Functional acrylate ester |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.